Anti-RHOT2 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A60872-50UL A60872-100UL
77717-339EA 360.84 CAD
77717-339 77717-338
Anti-RHOT2 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: RHOT2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR