Anti-COQ8B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44895-50UL A44895-100UL
77708-467EA 360.84 CAD
77708-467 77708-466
Anti-COQ8B Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: COQ8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR