Anti-VSIG10 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44894-100UL A44894-50UL
77708-464EA 529.22 CAD
77708-464 77708-465
Anti-VSIG10 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: VSIG10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR