Anti-CALCA Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A44853-50UG A44853-100UG
77708-389EA 360.84 CAD
77708-389 77708-388
Anti-CALCA Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR