Anti-CALCA Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44852-100UG A44852-50UG
77708-386EA 529.22 CAD
77708-386 77708-387
Anti-CALCA Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR