Anti-FGF18 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44815-50UL A44815-100UL
77708-317EA 360.84 CAD
77708-317 77708-316
Anti-FGF18 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: FGF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR