Anti-ALPL Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44800-100UG A44800-50UG
77689-154EA 529.22 CAD
77689-154 77708-290
Anti-ALPL Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: ALPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR