Anti-SLC16A7 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44793-50UL A44793-100UL
77708-278EA 360.84 CAD
77708-278 77708-277
Anti-SLC16A7 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SLC16A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR