Anti-GADD45A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44789-100UL A44789-50UL
77708-269EA 529.22 CAD
77708-269 77708-270
Anti-GADD45A Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: GADD45A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR