Anti-ZNF146 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44724-100UL A44724-50UL
77708-157EA 529.22 CAD
77708-157 77708-158
Anti-ZNF146 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: ZNF146
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR