Anti-MAGEA2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44666-50UL A44666-100UL
77708-052EA 360.84 CAD
77708-052 77708-051
Anti-MAGEA2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000

Type:
Antigen: MAGEA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR