Anti-MAPK14 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44600-100UG A44600-50UG
77705-315EA 400.93 CAD
77705-315 77707-940
Anti-MAPK14 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, ELISA:1:10000

Type:
Antigen: MAPK14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR