Anti-CUEDC2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44564-50UL A44564-100UL
77707-873EA 360.84 CAD
77707-873 77707-872
Anti-CUEDC2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: CUEDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR