Anti-MLN Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44382-100UG A44382-50UG
77707-558EA 529.22 CAD
77707-558 77707-559
Anti-MLN Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: MLN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR