Anti-RAB25 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A44376-50UL A44376-100UL
77707-550EA 360.84 CAD
77707-550 77707-549
Anti-RAB25 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RAB25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR