Anti-GREM2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44229-100UG A44229-50UG
77705-311EA 400.93 CAD
77705-311 77707-292
Anti-GREM2 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000,

Type:
Antigen: GREM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR