Anti-SDHA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44221-50UL A44221-100UL
77707-279EA 360.84 CAD
77707-279 77707-278
Anti-SDHA Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:30-1:150

Type:
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR