Anti-TERT Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A44192-50UG A44192-100UG
77707-223EA 360.84 CAD
77707-223 77707-222
Anti-TERT Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR