Anti-TERT Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A44191-100UG A44191-50UG
77707-220EA 529.22 CAD
77707-220 77707-221
Anti-TERT Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR