Anti-TERT Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A44190-50UG A44190-100UG
77707-219EA 360.84 CAD
77707-219 77707-218
Anti-TERT Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR