Anti-TERT Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44189-100UG A44189-50UG
77689-124EA 529.22 CAD
77689-124 77707-217
Anti-TERT Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TERT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR