Anti-MYOD1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44170-50UL A44170-100UL
77707-186EA 360.84 CAD
77707-186 77707-185
Anti-MYOD1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: MYOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR