Anti-RETN Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A44080-50UL A44080-100UL
77707-037EA 360.84 CAD
77707-037 77707-036
Anti-RETN Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RETN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR