Anti-RAB25 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A62468-100UL A62468-50UL
77720-292EA 529.22 CAD
77720-292 77720-293
Anti-RAB25 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:40-1:150

Type:
Antigen: RAB25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR