Anti-MPG Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62439-50UL A62439-100UL
77720-250EA 360.84 CAD
77720-250 77720-249
Anti-MPG Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000

Type:
Antigen: MPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR