Anti-FES Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62384-50UL A62384-100UL
77720-158EA 360.84 CAD
77720-158 77720-157
Anti-FES Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: FES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR