Anti-CDH4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62369-50UL A62369-100UL
77720-130EA 360.84 CAD
77720-130 77720-129
Anti-CDH4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: CDH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR