Anti-TRIM45 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62310-50UL A62310-100UL
77720-035EA 360.84 CAD
77720-035 77720-034
Anti-TRIM45 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: TRIM45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR