Anti-C2orf15 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62219-100UG A62219-50UG
77690-186EA 529.22 CAD
77690-186 77719-873
Anti-C2orf15 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200

Type:
Antigen: C2orf15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR