Anti-DLG5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62186-50UL A62186-100UL
77719-810EA 360.84 CAD
77719-810 77719-809
Anti-DLG5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:50-1:200

Type:
Antigen: DLG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR