Anti-DNAH3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62182-100UG A62182-50UG
77690-183EA 529.22 CAD
77690-183 77719-802
Anti-DNAH3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: DNAH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol DNAH3
Environmentally Preferable
Antigen synonyms Dynein heavy chain 3|Dnahc3-b|Ciliary dynein heavy chain 3|DYH3_HUMAN|HsADHC3|Axonemal beta dynein heavy chain 3|axonemal|Dnah3
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID Q8TD57
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Recombinant Human Dynein heavy chain 3, axonemal protein (1-150AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR