Anti-GSDMB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62158-100UG A62158-50UG
77690-179EA 529.22 CAD
77690-179 77719-758
Anti-GSDMB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200

Type:
Antigen: GSDMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR