Anti-TMPRSS3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62149-50UL A62149-100UL
77719-742EA 360.84 CAD
77719-742 77719-741
Anti-TMPRSS3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:25-1:100

Type:
Antigen: TMPRSS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR