Anti-CAT1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A61744-100UL A61744-50UL
77718-978EA 529.22 CAD
77718-978 77718-979
Anti-CAT1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CAT1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR