Anti-SORL1 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A61712-100UG A61712-50UG
77718-917EA 529.22 CAD
77718-917 77718-918
Anti-SORL1 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SORL1
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR