Anti-CPT1B Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A61693-50UG A61693-100UG
77718-882EA 360.84 CAD
77718-882 77718-881
Anti-CPT1B Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CPT1B
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR