Anti-SCFD1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63514-100UL A63514-50UL
77690-289EA 529.22 CAD
77690-289 77722-278
Anti-SCFD1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: SCFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR