Anti-SLC35F5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63505-100UG A63505-50UG
77690-287EA 529.22 CAD
77690-287 77722-262
Anti-SLC35F5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: SLC35F5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR