Anti-SRFBP1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A63414-100UG A63414-50UG
77722-090EA 529.22 CAD
77722-090 77722-091
Anti-SRFBP1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SRFBP1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol SRFBP1
Environmentally Preferable
Antigen synonyms SRFB1_HUMAN|SRFBP1|p49|p49/STRAP|Serum response factor-binding protein 1|serum response factor binding protein 1|SRFBP 1|p49 / STRAP|SRF-dependent transcription regulation-associated protein|SRF dependent transcription regulation associated protein
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Gene ID Q8NEF9
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Serum response factor-binding protein 1 protein (201-429AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR