Anti-C7orf31 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63381-100UG A63381-50UG
77690-276EA 529.22 CAD
77690-276 77722-026
Anti-C7orf31 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200

Type:
Antigen: C7orf31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR