Anti-NMNAT3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63323-100UG A63323-50UG
77690-270EA 529.22 CAD
77690-270 77721-916
Anti-NMNAT3 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: NMNAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR