Anti-CPNE2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A63211-50UG A63211-100UG
77721-703EA 360.84 CAD
77721-703 77721-702
Anti-CPNE2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CPNE2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR