Anti-AIDA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63159-50UL A63159-100UL
77721-606EA 360.84 CAD
77721-606 77721-605
Anti-AIDA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:50-1:200

Type:
Antigen: AIDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR