Anti-DEDD2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63058-100UL A63058-50UL
77721-407EA 529.22 CAD
77721-407 77721-408
Anti-DEDD2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: DEDD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR