Anti-SELENOM Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A63053-50UG A63053-100UG
77721-398EA 360.84 CAD
77721-398 77721-397
Anti-SELENOM Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SELENOM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR