Anti-PPTC7 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62939-50UG A62939-100UG
77721-181EA 360.84 CAD
77721-181 77721-180
Anti-PPTC7 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: PPTC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR