Anti-SLC30A7 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A62909-100UG A62909-50UG
77721-125EA 529.22 CAD
77721-125 77721-126
Anti-SLC30A7 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SLC30A7
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR