Anti-TRIB1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A62762-50UG A62762-100UG
77720-850EA 360.84 CAD
77720-850 77720-849
Anti-TRIB1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TRIB1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR