Anti-TRIB1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62760-100UG A62760-50UG
77690-217EA 529.22 CAD
77690-217 77720-846
Anti-TRIB1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:50-1:200

Type:
Antigen: TRIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR