Anti-CNDP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A62701-50UL A62701-100UL
77720-733EA 360.84 CAD
77720-733 77720-732
Anti-CNDP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200

Type:
Antigen: CNDP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR